Please login to your online account to display your discounted pricing

Molecular Model Sets


Illustrate firsthand the significance of chemical formulas.

Manufacturer: fisher science education  S440522

 View more versions of this product

Catalog No. S44052-2

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Includes Balls
Type Molecular model basic set replacement balls; 32mm
For Use With (Application) Visualize the 3D nature of a molecule Demonstrates organic, inorganic and molecular structure
Color Black (C)