Please login to your online account to display your discounted pricing

Molecular Model Sets


Illustrate firsthand the significance of chemical formulas.

Manufacturer: fisher science education  S65337

 View more versions of this product

Catalog No. S65337

  • / Pack
 

Description & Specifications

Specifications

Type Advanced set replacement balls molecular model; 20mm
Color White (H)
For Use With (Application) Visualize the 3D nature of a molecule Demonstrates organic, inorganic and molecular structure