Christensen Education Molecular Model Organic Structure Set
Organic structure model set allows you to clearly illustrate the significance of chemical formulas.
Manufacturer:  Christensen Education EPD9078
 View more versions of this product
Catalog No. S44052
Includes: Molecular model sets include balls, as indicated in chart; pegs; springs; and detailed instructions. Replacement bond set includes 1.25 in. (3.18cm), wooden pegs (30), 2 in. (5.1cm) wooden pegs (10), and 2.13 in. (6.5cm) springs (10). Replacement ball sets include balls only.
Description
Description
  • Allow students to visualize the three-dimensional nature of a molecule
  • Basic Set demonstrates organic structure
  • Advanced Set demonstrates organic, inorganic, and molecular structure
Specifications
Specifications
Visualize the 3D nature of a molecule Demonstrates organic, inorganic and molecular structure
1.25 in. dia. balls: 10 - black/Carbon tetrahedral (4 holes), 2 -Blue/Nitrogen (3 holes); 1.125 in. dia. balls: 28 - yellow/Hydrogen (1 hole), 6 -red/Oxygen (2 holes), 4 - green/Chlorine (1 hole), 2 - orange/Bromine (1 hole), 2 - Purple/Iodine or Fluorine; Bonds: 30 - 1.25 in. wooden pegs, 10 - 2.125 in. pegs, 10 - 2 in. springs.
Molecular model set