Thermo Scientific™ Molecular BioProducts™ SoftFit-L Reload™ System: Sterile Tips
Reload tip system provides space-saving and environmental benefits
$198.58 - $242.35
Specifications
For Use With (Equipment) Rainin LTS Pipetters
Sterility Sterile
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Packaging Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Model Packaging Volume (Metric) Price Quantity  
21-402-556 Thermo Scientific™
MB3721RI
Pure 20L Reload, 10 x 96 20μL Pack of 960 Each for $216.87
Case of 5 Pack for $1,084.35
21-402-557 Thermo Scientific™
MB372105RI
Pure 20L, Low Retention Reload, 10 x 96 20μL Pack of 960 Each for $235.19
Case of 5 Pack for $1,175.95
21-402-567 Thermo Scientific™
MB3751RI
Pure 200L Reload, 10 x 96 200μL Pack of 960 Each for $242.35
Case of 5 Pack for $1,211.75
21-402-568 Thermo Scientific™
MB375105RI
Pure 200L, Low Retention Reload, 10 x 96 200μL Pack of 960 Each for $235.19
Case of 5 Pack for $1,175.95
21-402-577 Thermo Scientific™
MB3731RI
Pure 300L Reload, 10 x 96 300μL Pack of 960 Each for $216.87
Case of 5 Pack for $1,084.35
21-402-578 Thermo Scientific™
MB373105RI
Pure 300L, Low Retention Reload, 10 x 96 300μL Pack of 960 Each for $216.87
Case of 5 Pack for $1,084.35
21-402-588 Thermo Scientific™
MB378105RI
Pure 1000L, Low Retention Reload, 8 x 96 1000μL Pack of 768 Each for $198.58
Case of 4 Pack for $794.32
21-402-587 Thermo Scientific™
MB3781RI
Pure 1000L Reload, 10 x 96 1000μL Pack of 768 Each for $198.58
Case of 4 Pack for $794.32
21-402-598 Thermo Scientific™
MB3741RI
Pure 1200L Reload, 8 x 96 1200μL Pack of 768 Each for $198.58
Case of 4 Pack for $794.32
Specifications
Description
The SoftFit-L Reload System is a complete product offering of pipet tips compatible with LTS™ pipetters made by Rainin Instrument, LLC. Designed in response to customer demands, this unique system provides preloaded pipet tips in color-coded inserts by volume size with the option of an easy-to-use “push down” reload pack. Choose sterile or nonsterile tips, in five volume sizes, with or without our proprietary low-retention technology. SoftFit tips are also available with MBP's patented ART™ barrier; contact your Fisher Scientific Customer Service Representative for more information.
Benefits
  • Certified RNase-, DNase- and Pyrogen-free
  • Recessed lid allows stacking of reloads and racks
  • Unique, easy-to-use push-down reload sleeve
  • Easy, one-hand operation of locking clasp Pipet tip plates clip securely into racks but are easy to remove
  • Part and lot numbers are printed on each insert for full traceability
  • Strong design prevents flexing and warping
  • Recessed holes stabilize pipet tips in multichannel applications
  • Fully recyclable components
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Rainin LTS Pipetters
Sterile