Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
$56.03 - $56.03
Specifications
Cap Style Flat
Certifications/Compliance RNase, DNase, RNA, DNA, ATP, pyrogen-free
Material Polypropylene
Graduated No
Includes Flat cap
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Sterile Price Quantity  
Catalog Number Mfr. No. Color Sterile Price Quantity  
21-403-182 Thermo Scientific™
MB3444B
Blue No Pack of 500 Each for $56.03
Case of 10 Pack for $560.30
21-403-184 Thermo Scientific™
MB3444G
Green No Pack of 500 Each for $56.03
Case of 10 Pack for $560.30
21-403-186 Thermo Scientific™
MB3444Y
Yellow No Pack of 500 Each for $56.03
Case of 10 Pack for $560.30
Specifications
Description
  • With flat cap
  • No frosting or graduations
  • Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Flat
Polypropylene
Flat cap
RNase, DNase, RNA, DNA, ATP, pyrogen-free
No
Bag