Please login to your online account to display your discounted pricing
Thermo Scientific™ Molecular BioProducts™ MBP™ and Pure™ Pipet Tips with SoftFit™ L Design
Compatible with Rainin™ LTS™ LiteTouch™ Pipetters
$193.55 - $258.45
Specifications
For Use With (Equipment) Rainin LTS Pipetters
Tip Style Rainin LTS
View More Specs
Products
Catalog Number Mfr. No. Packaging Sterility Volume (Metric) Price Quantity    
21-236-04
thermo scientific™
MB3752
Tray, 10 x 96 Nonsterile 200μL Pack of 960 Each for $212.03
Case of 5 Pack for $1,060.15
21-236-01
thermo scientific™
MB3721
10 x 96/Rack Sterile 20μL Pack of 960 Each for $247.19
Case of 5 Pack for $1,235.95
21-236-03
thermo scientific™
MB3751
10 x 96/Rack Sterile 200μL Pack of 960 Each for $247.19
Case of 5 Pack for $1,235.95
21-236-05
thermo scientific™
MB3781
Tray, 8 x 96 Sterile 1000μL Pack of 768 Each for $221.68
Case of 4 Pack for $886.72
21-236-06
thermo scientific™
MB3782
Tray, 10 x 96 Nonsterile 1000μL Pack of 768 Each for $193.55
Case of 4 Pack for $774.20
21-402-554
thermo scientific™
MB3722HR
Hinged Rack, 10 x 96 Nonsterile 20μL Pack of 960 Each for $212.03
Case of 5 Pack for $1,060.15
21-402-552
thermo scientific™
MB3721HR
Hinged Rack, 10 x 96 Sterile 20μL Pack of 960 Each for $247.19
Case of 5 Pack for $1,235.95
21-402-563
thermo scientific™
MB3751HR
Hinged Rack, 10 x 96 Sterile 200μL Pack of 960 Each for $247.19
Case of 5 Pack for $1,235.95
21-402-565
thermo scientific™
MB3752HR
Hinged Rack, 10 x 96 Nonsterile 200μL Pack of 960 Each for $212.03
Case of 5 Pack for $1,060.15
21-402-573
thermo scientific™
MB3731HR
Hinged Rack, 10 x 96 Sterile 300μL Pack of 960 Each for $258.45
Case of 5 Pack for $1,292.25
21-402-575
thermo scientific™
MB3732HR
Hinged Rack, 10 x 96 Nonsterile 300μL Pack of 960 Each for $212.03
Case of 5 Pack for $1,060.15
21-402-583
thermo scientific™
MB3781HR
Hinged Rack, 8 x 96 Sterile 1000μL Pack of 768 Each for $221.68
Case of 4 Pack for $886.72
21-402-585
thermo scientific™
MB3782HR
Hinged Rack, 8 x 96 Nonsterile 1000μL Pack of 768 Each for $193.55
Case of 4 Pack for $774.20
21-402-594
thermo scientific™
MB3741HR
Hinged Rack, 8 x 96 Sterile 1200μL Pack of 768 Each for $221.68
Case of 4 Pack for $886.72
21-402-596
thermo scientific™
MB3742HR
Hinged Rack, 8 x 96 Nonsterile 1200μL Pack of 768 Each for $193.55
Case of 4 Pack for $774.20
Description & Specifications
Specifications
For Use With (Equipment) Rainin LTS Pipetters
Tip Style Rainin LTS
SoftFit-L tips have thinner walls than conventional tips so they conform more easily to the pipetter barrel, which ensures a better seal and allows the tip to eject with less force.
  • ART Non-Filtered tips are available in sterile and non-sterile
  • Available racked or in hinged racks
Hinged Racks
  • Covers can be used as hinged or lift-off style
  • Easy, one-hand operation of locking clasp
  • Pipet tip inserts clip securely into racks but are easy to remove
  • Empty rack part numbers allow for easy conversion to Reload System
  • All components are completely recyclable and contain recycling symbols
  • Economical and environmental design reduces waste and saves bench space
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.