missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Moldex™ Special Ops Series Camo Earmuff

Camouflage color and a sleek, curvy shape

$355.08 - $355.08

Specifications

Color Camouflage
Product Type Ear Muffs
For Use With (Application) For protection in the work place
View More Specs

Products 1
Catalog Number Mfr. No. Noise Reduction Rating (NRR) Price Quantity  
Catalog Number Mfr. No. Noise Reduction Rating (NRR) Price Quantity  
19-160-0155
View Documents
Moldex™
6209
26 dB
Case of 20 Each for $355.08
 
Description

Description

Stylish protection in the work place.

  • Sleek, curvy shape
  • Camouflage color
  • 100% PVC free

M1 Series

  • Black-chrome, spring-steel headband resists fatigue and doesn't twist out of shape
  • Six different adjustment points provide a custom and comfortable fit
  • Folds into a compact size for easy storage
  • Noise Reduction Rating: 29
M2 Series
  • Special molded grips make fitting and alignment quick and easy
  • Cup adjustment is infinitely variable and distributes pressure evenly for a comfortable fit
  • Noise Reduction Rating: 26

Specifications

Specifications

Camouflage
Ear Muffs
For protection in the work place
SDS