Moldex™ 8000 Series Pre-Assembled Multi-Gas Vapor Smart™ Cartridge Respirators
Pre-assembled respirator kits simplify inventory and are ideal for use against multiple hazards in the same workplace. Includes 8000 Series facepiece and 8600 Multi-Gas/Vapor Smart Cartridges.
Supplier: Moldex™ 8602
Description
- 8000 Series half-mask pre-assembled with 8600 multi-gas/vapor smart cartridges
- Simplifies inventory by reducing need to stock multiple cartridge types
- Approved for nine common gas/vapors, replaces up to five gas/vapor cartridges
- Ideal for use against multiple hazards in same workplace
- For oil-free paint sprays and pesticides
- Soft, durable facepiece and lightweight design with extra-wide sealing
- Exhalation valve positioned for better communication
- Cartridges have a snap-in mount for easy replacement and no misthreading
- Living hinge allows head harness to be folded comfortably around neck when not in use
- Low profile, compact design fits well under welding helmets and with safety glasses
- Maintenance made simple with very few replacement parts
1 Includes two 19-003-346 N95 particulate prefilters, two 18-110 retainers, and two 18-102 organic vapor cartridges.
Specifications
Multi-Gas, Vapor Smart™ Cartridges | |
Gray | |
Pre-Assembled Half-Mask Respirator | |
Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing | |
8602 |
NIOSH 42 CFR Part 84, TC-23C-1223 | |
Thermoplastic Elastomer | |
Medium | |
8000 Facepiece, 8600 Multi-Gas, Vapor Smart™ Cartridges |