Moldex™ HandyStrap™ Series Particulate Respirators
Buckle allows mask to hang around neck when not in use
Manufacturer:  Moldex™ 2730AN100
 View more versions of this product
Catalog No. 19-150-0141
Description
Description
Disposable, maintenance-free design features convenient HandyStrap. Loosen the strap to allow the mask to hang freely around the neck when not in use
Application
  • See table matrix below
  • Design
    • HandyStrap feels cool and comfortable against skin
    • Strong Dura-Mesh™ shell resists collapsing in regular use and even under hot and humid conditions
    • Features comfortable contour-molded nose bridge with soft foam nose flange
    • Softspun™ lining enhances durability
    • 100% PVC free
    Styles
    • The 2600 N95 Series provides economical protection
    • All models (except 2600 N95) feature Ventex™ exhalation valve that reduces breathing resistance and vents hot, humid breath; workers breathe easier and stay cooler
    • The 2800 N95 and 2840 R95 Series include an added carbon layer to filter out nuisance levels of ozone and organic vapors (less than OSHA PEL)
    • The 2940 R95 Series protects against oil and oil-based particulates while filtering out nuisance levels of acid gas irritants (less than OSHA PEL)
    • Special Ops™ Models offer the same protection with a black Dura-Mesh shell
    Certifications
    All are NIOSH certified. N95: 95% efficient, not for oil; N100: 99.97% efficient, not for oil; R95: 95% efficient, resistant to oil.
    Compliance
    Meet heat and flame resistance requirements in accordance with ANSI™/ISEA 110-2003 Section 7.11.1.
    Specifications
    Specifications
    30 Case
    95% or greater against particulate aerosols free of oil
    Yes
    Pharmaceutical or battery manufacturing, welding and metal pouring
    Dura Mesh™
    Medium/Large
    Particulate Respirator
    White
    N100 Filter
    Ventex™ valve and handystrap™
    Non-oil based particles
    ANSI ISEA 110-2003 7.11.1., NIOSH
    Documents