Moldex™ 8920 Disk Piggyback Adapter
For use with 8000 series half mask respirators
Supplier: Moldex™ 8920
Description
Used to piggyback (8940) P100 filter with gas/vapor cartridgesSpecifications
Filter Cartridge | |
Adapter | |
Retainer ring |
NIOSH | |
P100 particulate filter disc in piggyback combination with 8000 series cartridge | |
6 Case |