Moldex™ EZ-On™ Particulate Respirators
The first and only NIOSH N95 approved adjustable one-strap respirator
Manufacturer:  Moldex™ EZ23S
 View more versions of this product
Catalog No. 19-149-722
Description
Description
This N95 respirator offers a quick fit for fast donning and doffing. Its one adjustable cloth strap with FLEXWING™ design feeds through a harness that secures the strap against the head and prevents slipping, snagging, and tangling.
Application
  • Hot, humid, and dusty environments where no oils are present, such as welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
  • Design
    • Dura-Mesh™ shell surrounds the mask's filter media, keeping it cleaner-looking longer; also helps respirator to hold its shape longer, extending service life
    • Design features no metal nose band to pinch closed; a soft foam nose cushion and contoured shape ensure a comfortable face seal
    • Available with or without a Ventex™ exhalation valve; valves help hot air to escape the mask faster, for cooler, more natural breathing
    Certifications
    NIOSH certified under 42CFR84 regulations to have a filter efficiency level of 95% or greater against non–oil-based particulate aerosols up to 10X the Permissible Exposure Limit (PEL).
    Specifications
    Specifications
    10 Pack
    95% or greater against non-oil based particulate aerosols
    Yes
    Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
    Dura Mesh™
    Small
    E-Z On™ Particulate Respirator
    White
    N95 Filter
    Ventex™ valve
    Aerosols free of oil
    NIOSH TC-84A-3746, N95
    Documents