Moldex™ Comets™ Reusable Ear Plugs
Comets™ reusable ear plugs feature a triple flange design that provides maximum comfort even when worn for long periods of time.
Manufacturer:  Moldex™ 6490
 View more versions of this product
Catalog No. 29-635-5749
Description
Description
  • User-friendly finger pocket makes insertion easy
  • Extra soft material and an air-cushioned tip provide all-day comfort
  • Soft triple flange adjusts to most size ear canals to hold the plug securely in place
  • 100% PVC-Free™
Specifications
Specifications
Yes
Universal
No
50 Pairs/Pk.
Green
Uncorded
Reusable
Class BL
Utilities, Manufacturing, Production, Mining, Oil and Gas, Transportation, and Other Various Industries
Poly bag
PVC-free
25 dB
Flanged
No