Moldex™ 8000 Series Half-Mask Facepieces
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
Supplier:  Moldex™ MDX8001
 View more versions of this product
Catalog No. 18-999-1029
Description
Description
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
  • Extra wide surface seals easily and stays sealed even when talking
  • Low profile design provides a large field of vision
  • Facepiece has no gaskets or yoke; fewer parts to maintain and inventory
  • Each cartridge includes a new inhalation valve and features snap-in mount that eliminates mis-threading
Specifications
Specifications
NIOSH
Thermoplastic Elastomer
Small
Snap-in gasket
Half Mask Facepiece