Please login to your online account to display your discounted pricing

Moldex™ 2360-P100 Particulate Respirator with Ventex™ Valve

All-day comfort with P100 protection

$69.75 - $69.75

Specifications

Certifications ANSI ISEA 110-2003 7.11.1.
Color White
Product Type Particulate Respirator
Filter Type P100 Filter
For Use With (Application) Environments where an osha substance specific standard applies (lead, cadmium, arsenic, mda) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

19-059-332

 
moldex™
2360P100
5 Pack of 5 Each for $69.75
Case of 4 Pack for $279.00
Description & Specifications

Specifications

Certifications ANSI ISEA 110-2003 7.11.1.
Color White
Product Type Particulate Respirator
Filter Type P100 Filter
For Use With (Application) Environments where an osha substance specific standard applies (lead, cadmium, arsenic, mda) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
Includes Ventex™ valve, soft foam nose cushion
Size Medium/Large

Application

  • Use in environments where an OSHA substance-specific standard applies (lead, cadmium, arsenic, MDA) as well as pharmaceutical manufacturing, conventional and nuclear power generation, welding, cutting and metal pouring
  • Design
    • Adjustable cloth straps assure snug, comfortable fit
    • Ventex valve lets the hot air out faster so workers breathe easier and stay cooler
    • Soft foam face cushion for added comfort and no pressure points
    • Dura-Mesh™ shell resists collapsing in regular use and even under hot and humid conditions
    • Contour-molded nose bridge seals without metal noseband
    • Single use

    Do not use against concentrations of particulates that exceed the maximum use concentration or 10X the OSHA PEL, whichever is lower.

    NIOSH certified to have a filter efficiency of 99.97% against oil and non-oil-based particulates. NIOSH Approval No.: TC-84A-3642.

    Meets heat and flame resistance requirements in accordance with ANSI™/ISEA 110-2003 Section 7.11.1.