Please login to your online account to display your discounted pricing
Mohs Hardness Scale
Learn how hardness relates to mineral identification.
Manufacturer: fisher science education  AZ-20-3941
 View more versions of this product
Catalog No. S90622
  • / Each
Description & Specifications
Specifications
Product Type Mohs hardness scale kit; 1 Ea.
For Use With (Application) Used to classify minerals by hardness
Packaging Quantity 1
Includes Card-mounted mineral specimens and additional activities