Mohs Hardness Scale
Learn how hardness relates to mineral identification.
$40.40 - $40.40
Specifications
For Use With (Application) Used to classify minerals by hardness
Product Type Mohs hardness scale kit; 1 Ea.
Includes Card-mounted mineral specimens and additional activities
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
S90622 Fisher Science Education
AZ-20-3941
1 Each for $40.40
Specifications
Description
  • Developed more than 200 years ago by Friedrich Mohs
  • Used to classify minerals by hardness on a scale of 1 to 10
  • Determine and study the hardness of included minerals
  • Card-mounted mineral specimens reference hardness for scale values 1 through 9
  • Additional activities allow testing for other key mineral properties
Description & Specifications
Specifications
Used to classify minerals by hardness
Mohs hardness scale kit; 1 Ea.
Card-mounted mineral specimens and additional activities