Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Moesin Ab-1, Mouse Monoclonal Antibody


Deliver accurate, reproducible results with the Thermo Scientific™ Moesin Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS727P1ABX

 View more versions of this product

Catalog No. MS727P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Rat
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Moesin purified from cow uterus
Quantity 100µL
Format Concentrated
Isotype IgG2a kappa
Regulatory Status RUO
Clone 38/87
Antigen Moesin Ab-1
Host Species Mouse
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Cytoskeletal Protein
Concentration 1mg/mL

Deliver accurate, reproducible results with the Thermo Scientific™ Moesin Ab-1, Mouse Monoclonal Antibody.

Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.

Host Species: Mouse

Clone: 38/87

Isotype: IgG1/κ

Species Reactivity: Human, Cow and Rat. Others not known.

Epitope: Not determined

Immunogen: Moesin purified from cow uterus

Molecular Weight: 78kDa moesin and 80kDa radixin

Positive Control: Raji cells; Placenta

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured only)
  • Inhibits (40%) Binding of Proteoheparan to Smooth Muscle Cells
  • Inhibits Proliferation of Smooth Muscle Cells

Note:

USA: RUO; Int'l: RUO