Please login to your online account to display your discounted pricing
Model Steam Engine
Use this steam engine to observe energy conversions
$311.85 - $311.85
Specifications
Size (Metric) Bore 1.27cm
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S60675
fisher science education
75
Model 75 Dry Fuel Each for $311.85
Description & Specifications
Specifications
Size (Metric) Bore 1.27cm
Model burns a fuel pellet, heating water to steam, which moves the piston and drives the flywheel.
Supplied with 20 fuel pellets. Electrical model is also available.