Please login to your online account to display your discounted pricing
Model 3000 Sand, Gravel, and Bedrock Groundwater Flow Simulator
Plastic bedrock layer accurately demonstrates flow in a fractured/cavernous aquifer.
Manufacturer: fisher scientific  E3000OPT4
 View more versions of this product
Catalog No. S92501D
  • / Each
Description & Specifications
Specifications
Material Acrylic
For Use With (Application) To demonstrate a complete water cycle, by recycling the water via a pump mechanism
Includes Mini pump, instruction manual, siphon bottle, syringe pumping assembly, three pipets, three neon gimp cords or three extra-large neon Rubber bands, valve turner, four colors of dye, drain tube and rainmaker in ultra carrying case
Product Type Bedrock Simulator