Please login to your online account to display your discounted pricing
Model 3000 Sand, Gravel, and Bedrock Groundwater Flow Simulator
Plastic bedrock layer accurately demonstrates flow in a fractured/cavernous aquifer.
Manufacturer: fisher scientific  E3000OPT1
 View all options for this product
Catalog No. S92501A
Description & Specifications
For Use With (Application) To demonstrate a complete water cycle, by recycling the water via a pump mechanism
Product Type Bedrock Simulator
Includes Mini pump, instruction manual, siphon bottle, syringe pumping assembly, three pipets, three neon gimp cords or three extra-large neon Rubber bands, valve turner, four colors of dye, drain tube and carrying case
Material Acrylic
  • Similar to the Model 2000 Simulator (S92500A), plus a bedrock aquifer
  • Shows both unconfined and confined sand and gravel aquifers overlying cavernous limestone and fractured-bedrock aquifers
  • Particularly useful where local geology contains caves, sinkholes, or fractured nonporous bedrock
  • Demonstrations also relate to areas that have been extensively deep-mined
  • Fully assembled
  • S92501D includes simulator and Rainmaker in ultra case
  • Dimensions: 19.5 × 8 × 12 in. (49.5 × 20.3 × 30.5cm)