Model 3000 Sand, Gravel, and Bedrock Groundwater Flow Simulator
Plastic bedrock layer accurately demonstrates flow in a fractured/cavernous aquifer.
$4,374.00 - $4,374.00
Specifications
For Use With (Application) To demonstrate a complete water cycle, by recycling the water via a pump mechanism
Product Type Bedrock Simulator
Material Acrylic
View More Specs
Includes: Mini-pump, instruction manual, siphon bottle, syringe pumping assembly, three pipets, three neon gimp cords or three extra-large neon rubber bands, valve turner, four colors of dye, drain tube and carrying case
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
S92501D Fisher Scientific
E3000OPT4
Mini pump, instruction manual, siphon bottle, syringe pumping assembly, three pipets, three neon gimp cords or three extra-large neon Rubber bands, valve turner, four colors of dye, drain tube and rainmaker in ultra carrying case Each for $4,374.00
Specifications
Description
  • Similar to the Model 2000 Simulator (S92500A), plus a bedrock aquifer
  • Shows both unconfined and confined sand and gravel aquifers overlying cavernous limestone and fractured-bedrock aquifers
  • Particularly useful where local geology contains caves, sinkholes, or fractured nonporous bedrock
  • Demonstrations also relate to areas that have been extensively deep-mined
  • Fully assembled
  • S92501D includes simulator and Rainmaker in ultra case
  • Dimensions: 19.5 × 8 × 12 in. (49.5 × 20.3 × 30.5cm)
Description & Specifications
Specifications
To demonstrate a complete water cycle, by recycling the water via a pump mechanism
Bedrock Simulator
Acrylic