missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOBS, 0.2M buffer soln., pH 8.0

$79.11 - $124.00

Chemical Identifiers

CAS 115724-21-5
Molecular Formula C8H17NO4S
Molecular Weight (g/mol) 223.287
MDL Number MFCD00674916
InChI Key VTOWJTPBPWTSMK-UHFFFAOYSA-N
Synonym 4-(4-Morpholinyl)butanesulfonic acid
PubChem CID 3674064
IUPAC Name 4-morpholin-4-ylbutane-1-sulfonic acid
SMILES C1COCCN1CCCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63594AE View Documents Alfa Aesar™
J63594AE
100mL Each for $79.11
 
AAJ63594AK View Documents Alfa Aesar™
J63594AK
250mL Each for $124.00
 
Specifications

Chemical Identifiers

115724-21-5
223.287
VTOWJTPBPWTSMK-UHFFFAOYSA-N
3674064
C1COCCN1CCCCS(=O)(=O)O
C8H17NO4S
MFCD00674916
4-(4-Morpholinyl)butanesulfonic acid
4-morpholin-4-ylbutane-1-sulfonic acid

Specifications

115724-21-5
MFCD00674916
4-(4-Morpholinyl)butanesulfonic acid
C1COCCN1CCCCS(=O)(=O)O
223.287
Liquid
Colorless
MOBS buffer soln.
C8H17NO4S
0.2M buffer soln.
VTOWJTPBPWTSMK-UHFFFAOYSA-N
4-morpholin-4-ylbutane-1-sulfonic acid
3674064
8.0
100mL
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C