missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MOBS, 0.2M buffer soln., pH 7.5

$86.68 - $135.70

Chemical Identifiers

CAS 115724-21-5
Molecular Formula C8H17NO4S
Molecular Weight (g/mol) 223.287
MDL Number MFCD00674916
InChI Key VTOWJTPBPWTSMK-UHFFFAOYSA-N
Synonym 4-(4-Morpholinyl)butanesulfonic acid
PubChem CID 3674064
IUPAC Name 4-morpholin-4-ylbutane-1-sulfonic acid
SMILES C1COCCN1CCCCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63519AE View Documents Alfa Aesar™
J63519AE
100mL Each for $86.68
 
AAJ63519AK View Documents Alfa Aesar™
J63519AK
250mL Each for $135.70
 
Description

Description

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. Usage above 20 mM in mammalian cell culture work is not recommended.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

115724-21-5
223.287
VTOWJTPBPWTSMK-UHFFFAOYSA-N
3674064
C1COCCN1CCCCS(=O)(=O)O
C8H17NO4S
MFCD00674916
4-(4-Morpholinyl)butanesulfonic acid
4-morpholin-4-ylbutane-1-sulfonic acid

Specifications

115724-21-5
MFCD00674916
4-(4-Morpholinyl)butanesulfonic acid
C1COCCN1CCCCS(=O)(=O)O
223.287
Liquid
100mL
C8H17NO4S
0.2M buffer soln.
VTOWJTPBPWTSMK-UHFFFAOYSA-N
4-morpholin-4-ylbutane-1-sulfonic acid
3674064
7.5
MOBS buffer soln.