missing translation for 'onlineSavingsMsg'
Learn More

Mopec Mobile 2-Door Cabinet

Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.

$17467.68 - $19835.55

Specifications

Depth (English) 30 in.,30 in.
View More Specs

Products 2
Catalog Number Mfr. No. Depth (English) Price Quantity  
Catalog Number Mfr. No. Depth (English) Price Quantity  
21-381-143
View Documents
Mopec
LC106330
30 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
21-381-144
View Documents
Mopec
LC106336
36 in. N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Construction

  • Mobile Cabinet
  • (2) solid steel inset doors
  • (4) casters: (2) swivel, (2) fixed with locks
  • Includes finished top & finished back, interlocking drawers and anti-tilt weight

Materials

  • 304 stainless steel construction with a polished No. 4 finish

Dimensions

  • LC1063-30: 30 in. L x 22 in. W x 27.1 in. H
  • LC1063-36: 36 in. L x 22 in. W x 27.1 in. H
Specifications

Specifications

30 in.,30 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'