Please login to your online account to display your discounted pricing

MNI-caged-L-glutamate, Tocris

Stable photoreleaser of L-glutamate

$261.58 - $1,075.94

Chemical Identifiers

Chemical Name or Material MNI-caged-L-glutamate
Assay Percent Range >98%
Molecular Formula C14H17N3O6
Formula Weight 323.3
Synonym (S)-α-Amino-2, 3-dihydro-4-methoxy-7-nitro-δ-oxo-1H-indole-1-pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

149050

 
r & d systems
149050
50mg Each for $1,075.94

149010

 
r & d systems
149010
10mg Each for $261.58
Description & Specifications

Specifications

Formula Weight 323.3
Quantity 50mg
Chemical Name or Material MNI-caged-L-glutamate
Assay Percent Range >98%
Molecular Formula C14H17N3O6
CAS 295325-62-1
Synonym (S)-α-Amino-2, 3-dihydro-4-methoxy-7-nitro-δ-oxo-1H-indole-1-pentanoic acid
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C