Please login to your online account to display your discounted pricing

MNI-caged-D-aspartate, Tocris

Caged D-aspartate; NMDA agonist

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material MNI-caged-D-aspartate
Assay Percent Range >98%
Molecular Formula C13H15N3O6
Formula Weight 309.27
Synonym (R)-α-Amino-2, 3-dihydro-4-methoxy-7-nitro-γ-oxo-1H-indole-1-butanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

227750

 
r & d systems
RDS227750
50mg Each for $1,116.26

227710

 
r & d systems
RDS227710
10mg Each for $265.15
Description & Specifications

Specifications

Formula Weight 309.27
Quantity 50mg
Assay Percent Range >98%
Synonym (R)-α-Amino-2, 3-dihydro-4-methoxy-7-nitro-γ-oxo-1H-indole-1-butanoic acid
CAS 845555-94-4
Molecular Formula C13H15N3O6
Chemical Name or Material MNI-caged-D-aspartate
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C