missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MMP Inhibitor II

A potent, reversible, broad range inhibitor of matrix metalloproteinases

$206.85 - $336.54

Specifications

CAS 203915-59-7
Formula Weight 513.58g/mol
Synonym N-Hydroxy-1,3-bis(4-methoxyphenylsulfonyl)-5,5-dimethylhexahydropyrimidine-2-carboxamide
Storage Requirements −30° to −10°C
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65638MA
View Documents
Alfa Aesar™
J65638MA
10mg Each for $336.54
 
AAJ65638LB0
View Documents
Alfa Aesar™
J65638LB0
5mg Each for $206.85
 
Specifications

Specifications

203915-59-7
N-Hydroxy-1,3-bis(4-methoxyphenylsulfonyl)-5,5-dimethylhexahydropyrimidine-2-carboxamide
Solid
Off-white
Soluble in DMSO
513.58g/mol
−30° to −10°C
A potent, reversible, broad range inhibitor of matrix metalloproteinases
Signal Transduction Reagent