missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV), Rabbit Polyclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV), Rabbit Polyclonal Antibody.

$243.18 - $734.53

Specifications

Antigen MMP-9 (92kDa Collagenase IV)
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Host Species Rabbit
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB9234P0 Thermo Scientific™ Lab Vision™
RB9234P0
Purified Ab with BSA and Azide 100μL Each for $243.18
RB9234P1 Thermo Scientific™ Lab Vision™
RB9234P1
Purified Ab with BSA and Azide 500μL Each for $734.53
RB9234R7 Thermo Scientific™ Lab Vision™
RB9234R7
Ready-to-use for IHC (UVLP) 7mL Each for $482.45
Specifications

Description

Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV), Rabbit Polyclonal Antibody.

Metallomatrix proteases (MMPs) are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. This antibody reacts with both latent and active MMP-9.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Guinea Pig. Does not react with mouse, rat or bovine. Others not tested.

Epitope: Near C-terminus

Immunogen: A synthetic peptide derived from near C-terminal of human MMP-9 protein

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Placenta, or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

MMP-9 (92kDa Collagenase IV)
Polyclonal
Rabbit
Proteases And Inhibitors
A synthetic peptide derived from near C-terminal of human MMP-9 protein
Western Blot
Unlabeled
RUO
Primary
IgG