Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Ab-9, Rabbit Polyclonal Antibody

Deliver accurate, reproducible resultsThermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-9, Rabbit Polyclonal Antibody.

$272.76 - $1,379.02

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-9
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1539P

 
thermo scientific™ lab vision™
RB1539P
Purified Ab with BSA and Azide 1mL Each for $1,269.04

RB1539P0

 
thermo scientific™ lab vision™
RB1539P0
Purified Ab with BSA and Azide 100µL Each for $272.76

RB1539P1

 
thermo scientific™ lab vision™
RB1539P1
Purified Ab with BSA and Azide 500µL Each for $754.47

RB1539P1ABX

 
thermo scientific™ lab vision™
RB1539P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $843.69

RB1539PABX

 
thermo scientific™ lab vision™
RB1539PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,379.02
Description & Specifications

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-9
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Guinea Pig
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Type Primary
Immunogen A synthetic peptide from the middle region of human MMP-9
Isotype IgG

Deliver accurate, reproducible resultsThermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-9, Rabbit Polyclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Guinea Pig. Does not react with mouse, rat and cow. Others not known.

Epitope: Middle region of MMP-9

Immunogen: A synthetic peptide from the middle region of human MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned serum medium from (TPA-treated) HT-1080 cells; Placenta or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO