Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Ab-9, Rabbit Polyclonal Antibody


Deliver accurate, reproducible resultsThermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-9, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1539P1ABX

 View more versions of this product

Catalog No. RB1539P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 500µL
Isotype IgG
Regulatory Status RUO
Immunogen A synthetic peptide from the middle region of human MMP-9
Cross Reactivity Guinea Pig
Cross Reactivity Human
Antigen MMP-9 (92kDa Collagenase IV) Ab-9
Applications Western Blotting
Host Species Rabbit
Format Concentrated
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Classification Polyclonal
Type Primary
Concentration 1mg/mL
Item Description 500uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Deliver accurate, reproducible resultsThermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-9, Rabbit Polyclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Guinea Pig. Does not react with mouse, rat and cow. Others not known.

Epitope: Middle region of MMP-9

Immunogen: A synthetic peptide from the middle region of human MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned serum medium from (TPA-treated) HT-1080 cells; Placenta or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO