Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody.

$283.65 - $1,586.16

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-5
Clone IIA5
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS817B

 
thermo scientific™ lab vision™
MS817B
200μg/mL Biotin 1mL Each for $1,586.16

MS817B0

 
thermo scientific™ lab vision™
MS817B0
200μg/mL Biotin 100µL Each for $363.57

MS817B1

 
thermo scientific™ lab vision™
MS817B1
200μg/mL Biotin 500µL Each for $1,000.18

MS817P

 
thermo scientific™ lab vision™
MS817P
200μg/mL Unlabeled 1mL Each for $1,257.67

MS817P0

 
thermo scientific™ lab vision™
MS817P0
200μg/mL Unlabeled 100µL Each for $283.65

MS817P1

 
thermo scientific™ lab vision™
MS817P1
200μg/mL Unlabeled 500µL Each for $767.06

MS817P1ABX

 
thermo scientific™ lab vision™
MS817P1ABX
1mg/mL Unlabeled 100µL Each for $831.49
Description & Specifications

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-5
Clone IIA5
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen Human native 92kDa MMP-9
Isotype IgG2a kappa

Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IIA5

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells or CCL4 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not optimum)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO