Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody


Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS817P1ABX

 View more versions of this product

Catalog No. MS817P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rat
Immunogen Human native 92kDa MMP-9
Regulatory Status RUO
Antigen MMP-9 (92kDa Collagenase IV) Ab-5
Isotype IgG2a kappa
Applications Immunoprecipitation
Applications Western Blotting
Format Concentrated
Quantity 100µL
Clone IIA5
Conjugate Unlabeled
Concentration 1mg/mL
Classification Monoclonal
Type Primary
Research Discipline Proteases And Inhibitors

Deliver accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-5, Mouse Monoclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IIA5

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells or CCL4 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not optimum)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO