Please login to your online account to display your discounted pricing

Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody

Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody.

$272.78 - $1,391.04

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-4
Clone IA5
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS816P

 
thermo scientific™
MS816P
200μg/mL 1mL Each for $1,276.51

MS816P0

 
thermo scientific™
MS816P0
200μg/mL 100µL Each for $272.78

MS816P1

 
thermo scientific™
MS816P1
200μg/mL 500µL Each for $763.55

MS816P1ABX

 
thermo scientific™
MS816P1ABX
1mg/mL 100µL Each for $857.78

MS816PABX

 
thermo scientific™
MS816PABX
1mg/mL 200µL Each for $1,391.04
Description & Specifications

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-4
Clone IA5
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen Human native 92kDa MMP-9
Isotype IgG2a kappa

Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IA5

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO