Please login to your online account to display your discounted pricing

Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody


Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™  MS816P1ABX

 View more versions of this product

Catalog No. MS816P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Quantity 100µL
Format Concentrated
Cross Reactivity Human
Antigen MMP-9 (92kDa Collagenase IV) Ab-4
Isotype IgG2a kappa
Regulatory Status RUO
Immunogen Human native 92kDa MMP-9
Clone IA5
Host Species Mouse
Research Discipline Proteases And Inhibitors
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal

Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IA5

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO