Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody

Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody.

$279.47 - $1,409.88

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-1
Clone GE-213
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Immunohistochemistry (Frozen)
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS569P

 
thermo scientific™ lab vision™
MS569P
200μg/mL 1mL Each for $1,307.63

MS569P0

 
thermo scientific™ lab vision™
MS569P0
200μg/mL 100µL Each for $279.47

MS569P1

 
thermo scientific™ lab vision™
MS569P1
200μg/mL 500µL Each for $783.08

MS569P1ABX

 
thermo scientific™ lab vision™
MS569P1ABX
1mg/mL 100µL Each for $854.92

MS569PABX

 
thermo scientific™ lab vision™
MS569PABX
1mg/mL 200µL Each for $1,409.88
Description & Specifications

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-1
Clone GE-213
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen Purified human MMP-9
Isotype IgG1

Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: GE-213

Isotype: IgG1

Species Reactivity: Human. Others not known.

Immunogen: Purified human MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen tissues)
  • Immunoprecipitation (Native verified)
  • Inhibition of MMP-9 Activity

Note:

USA: RUO; Int'l: RUO

General References:

1. Nikkari ST, et. al. American Journal of Pathology, 1996, 149(5):1427-33.

2. Visscher DW, et. al. International Journal of Cancer, 1994, 59(3):339-44.

3. Schnaper HW, et. al. Journal of Cellular Physiology, 1993, 156(2):235-46.

4. Soini Y, et. al. Journal of Histochemistry and Cytochemistry, 1994, 42(7):945-51.

5. Hurskainen T, et. al. Journal of Histochemistry and Cytochemistry, 1996, 44(12):1379-88.