Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody


Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS569P1ABX

 View more versions of this product

Catalog No. MS569P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Quantity 100µL
Cross Reactivity Human
Antigen MMP-9 (92kDa Collagenase IV) Ab-1
Format Concentrated
Host Species Mouse
Immunogen Purified human MMP-9
Isotype IgG1
Regulatory Status RUO
Clone GE-213
Research Discipline Proteases And Inhibitors
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Supply accurate, reproducible results with the Thermo Scientific™ MMP-9 (92kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: GE-213

Isotype: IgG1

Species Reactivity: Human. Others not known.

Immunogen: Purified human MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen tissues)
  • Immunoprecipitation (Native verified)
  • Inhibition of MMP-9 Activity

Note:

USA: RUO; Int'l: RUO