missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ MMP-8 Substrate, fluorogenic

For cellular and molecular biology applications

Specifications

Substrate Type Fluorogenic
Storage Requirements -30° to -10°C
Color White to off-white
Molecular Weight 1042.10
Format Lyophilized
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
AAJ66790EXD Alfa Aesar™
J66790EXD
0.5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66790MCR Alfa Aesar™
J66790MCR
1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

Fluorogenic
White to off-white
Lyophilized
-30° to -10°C
1042.10