Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-7 (Matrilysin) Ab-1, Mouse Monoclonal Antibody

Supply accurate, reproducible results with the Thermo Scientific™ MMP-7 (Matrilysin) Ab-1, Mouse Monoclonal Antibody.

$278.33 - $1,415.53

Specifications

Antigen MMP-7 (Matrilysin) Ab-1
Clone ID2
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS813P

 
thermo scientific™ lab vision™
MS813P
200μg/mL 1mL Each for $1,303.24

MS813P0

 
thermo scientific™ lab vision™
MS813P0
200μg/mL 100μL Each for $278.33

MS813P1

 
thermo scientific™ lab vision™
MS813P1
200μg/mL 500μL Each for $783.08

MS813P1ABX

 
thermo scientific™ lab vision™
MS813P1ABX
1mg/mL 100μL Each for $861.20

MS813PABX

 
thermo scientific™ lab vision™
MS813PABX
1mg/mL 200μL Each for $1,415.53
Description & Specifications

Specifications

Antigen MMP-7 (Matrilysin) Ab-1
Clone ID2
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen Recombinant human matrilysin (PUMP-1)
Isotype IgG2b kappa

Supply accurate, reproducible results with the Thermo Scientific™ MMP-7 (Matrilysin) Ab-1, Mouse Monoclonal Antibody.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. The matrix metalloproteinase, matrilysin/MMP-7/PUMP-1, is thought to play an important role in the early steps of tumor progression. Expression of MMP2, -7, and -11 is greater in pancreatic carcinoma than in normal pancreas.

Host Species: Mouse

Clone: ID2

Isotype: IgG2b/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human matrilysin (PUMP-1)

Molecular Weight: ∽28kDa (proform) and ∽18kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human SW620 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO