Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-7 (Matrilysin) Ab-1, Mouse Monoclonal Antibody


Supply accurate, reproducible results with the Thermo Scientific™ MMP-7 (Matrilysin) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS813P1ABX

 View more versions of this product

Catalog No. MS813P1ABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Applications Immunofluorescence
Quantity 100µL
Regulatory Status RUO
Immunogen Recombinant human matrilysin (PUMP-1)
Host Species Mouse
Clone ID2
Cross Reactivity Human
Antigen MMP-7 (Matrilysin) Ab-1
Isotype IgG2b kappa
Conjugate Unlabeled
Research Discipline Proteases And Inhibitors
Type Primary
Classification Monoclonal
Concentration 1mg/mL

Supply accurate, reproducible results with the Thermo Scientific™ MMP-7 (Matrilysin) Ab-1, Mouse Monoclonal Antibody.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. The matrix metalloproteinase, matrilysin/MMP-7/PUMP-1, is thought to play an important role in the early steps of tumor progression. Expression of MMP2, -7, and -11 is greater in pancreatic carcinoma than in normal pancreas.

Host Species: Mouse

Clone: ID2

Isotype: IgG2b/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human matrilysin (PUMP-1)

Molecular Weight: ∽28kDa (proform) and ∽18kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human SW620 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting (Not verified)
  • Immunofluorescence
  • Affinity Purification

Note:

USA: RUO; Int'l: RUO