Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-3 (Stromelysin-1) Ab-2, Mouse Monoclonal Antibody

Produce accurate, reproducible results with the Thermo Scientific™ MMP-3 (Stromelysin-1) Ab-2, Mouse Monoclonal Antibody.

$271.55 - $1,573.06

Specifications

Antigen MMP-3 (Stromelysin-1) Ab-2
Clone SL-1 IID4
Applications Immunofluorescence
Applications Western Blotting
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS810B

 
thermo scientific™ lab vision™
MS810B
200μg/mL Biotin 1mL Each for $1,573.06

MS810B0

 
thermo scientific™ lab vision™
MS810B0
200μg/mL Biotin 100µL Each for $360.39

MS810B1

 
thermo scientific™ lab vision™
MS810B1
200μg/mL Biotin 500µL Each for $981.61

MS810P

 
thermo scientific™ lab vision™
MS810P
200μg/mL Unlabeled 1mL Each for $1,260.88

MS810P0

 
thermo scientific™ lab vision™
MS810P0
200μg/mL Unlabeled 100µL Each for $271.55

MS810P1

 
thermo scientific™ lab vision™
MS810P1
200μg/mL Unlabeled 500µL Each for $749.55

MS810P1ABX

 
thermo scientific™ lab vision™
MS810P1ABX
1mg/mL Unlabeled 100µL Each for $824.35
Description & Specifications

Specifications

Antigen MMP-3 (Stromelysin-1) Ab-2
Clone SL-1 IID4
Applications Immunofluorescence
Applications Western Blotting
Applications Immunofluorescence
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
Isotype IgG2b

Produce accurate, reproducible results with the Thermo Scientific™ MMP-3 (Stromelysin-1) Ab-2, Mouse Monoclonal Antibody.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Stromelysin (MMP3/Transin) degrades several matrix components including the core proteins of proteoglycans, laminin and non-helical regions of collagens.

Host Species: Mouse

Clone: SL-1 IID4

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: APMA (4-Aminophenylmercuric acetate) activated human stromelysin-1 (SL-1)

Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)

Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO