Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-3 (Stromelysin-1) Ab-2, Mouse Monoclonal Antibody


Produce accurate, reproducible results with the Thermo Scientific™ MMP-3 (Stromelysin-1) Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS810P1ABX

 View more versions of this product

Catalog No. MS810P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Applications Immunofluorescence
Applications Western Blotting
Format Concentrated
Clone SL-1 IID4
Antigen MMP-3 (Stromelysin-1) Ab-2
Cross Reactivity Human
Immunogen APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
Host Species Mouse
Isotype IgG2b
Quantity 100µL
Conjugate Unlabeled
Concentration 1mg/mL
Classification Monoclonal
Type Primary
Research Discipline Proteases And Inhibitors

Produce accurate, reproducible results with the Thermo Scientific™ MMP-3 (Stromelysin-1) Ab-2, Mouse Monoclonal Antibody.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Stromelysin (MMP3/Transin) degrades several matrix components including the core proteins of proteoglycans, laminin and non-helical regions of collagens.

Host Species: Mouse

Clone: SL-1 IID4

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: APMA (4-Aminophenylmercuric acetate) activated human stromelysin-1 (SL-1)

Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)

Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO