Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-2 (72kDa Collagenase IV) Ab-7, Rabbit Polyclonal Antibody

Produce accurate, reproducible results with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-7, Rabbit Polyclonal Antibody.

$274.45 - $1,354.65

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-7
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1537P

 
thermo scientific™ lab vision™
RB1537P
Purified Ab with BSA and Azide 1mL Each for $1,270.22

RB1537P0

 
thermo scientific™ lab vision™
RB1537P0
Purified Ab with BSA and Azide 100μL Each for $274.45

RB1537P1

 
thermo scientific™ lab vision™
RB1537P1
Purified Ab with BSA and Azide 500μL Each for $754.53

RB1537P1ABX

 
thermo scientific™ lab vision™
RB1537P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $844.49

RB1537PABX

 
thermo scientific™ lab vision™
RB1537PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,354.65
Description & Specifications

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-7
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen A synthetic peptide from the second half of human MMP-2
Isotype IgG

Produce accurate, reproducible results with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-7, Rabbit Polyclonal Antibody.

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat and Cow. Others not known.

Epitope: Second half of MMP-2

Immunogen: A synthetic peptide from the second half of human MMP-2

Molecular Weight: 72kDa (proform) and ∽66kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFl-1 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO