Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-2 (72kDa Collagenase IV) Ab-7, Rabbit Polyclonal Antibody


Produce accurate, reproducible results with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-7, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1537PABX

 View more versions of this product

Catalog No. RB1537PABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Isotype IgG
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Antigen MMP-2 (72kDa Collagenase IV) Ab-7
Quantity 1mL
Immunogen A synthetic peptide from the second half of human MMP-2
Applications Western Blotting
Host Species Rabbit
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Proteases And Inhibitors
Type Primary
Concentration 1mg/mL
Classification Polyclonal
Conjugate Unlabeled
Item Description 1mL; 1mg/mL; Unlabeled; Purified without BSA and azide

Produce accurate, reproducible results with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-7, Rabbit Polyclonal Antibody.

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat and Cow. Others not known.

Epitope: Second half of MMP-2

Immunogen: A synthetic peptide from the second half of human MMP-2

Molecular Weight: 72kDa (proform) and ∽66kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFl-1 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO