Please login to your online account to display your discounted pricing

Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody.

$272.88 - $1,605.41

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-4
Clone A-Gel VC2
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS806B

 
thermo scientific™
MS806B
200μg/mL Biotin 1mL Each for $1,605.41

MS806B0

 
thermo scientific™
MS806B0
200μg/mL Biotin 100μL Each for $368.10

MS806B1

 
thermo scientific™
MS806B1
200μg/mL Biotin 500μL Each for $1,001.59

MS806P

 
thermo scientific™
MS806P
200μg/mL Unlabeled 1mL Each for $1,290.49

MS806P0

 
thermo scientific™
MS806P0
200μg/mL Unlabeled 100μL Each for $272.88

MS806P1

 
thermo scientific™
MS806P1
200μg/mL Unlabeled 500μL Each for $768.86

MS806P1ABX

 
thermo scientific™
MS806P1ABX
1mg/mL Unlabeled 100μL Each for $856.24

MS806PABX

 
thermo scientific™
MS806PABX
1mg/mL Unlabeled 200μL Each for $1,398.24
Description & Specifications

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-4
Clone A-Gel VC2
Classification Monoclonal
Cross Reactivity Human
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen Human APMA-activated native 72kDa MMP-2
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody.

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.

Host Species: Mouse

Clone: A-Gel VC2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human APMA-activated native 72kDa MMP-2

Molecular Weight: 72kDa (proform) and ∽66kDa (active form)

Positive Control: Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (not qualified)

Note:

USA: RUO; Int'l: RUO