Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-2 (72kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody.

$270.71 - $1,581.78

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-1
Clone CA-4001 or CA719E3C
Applications Western Blot
Classification Monoclonal
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS567B

 
thermo scientific™ lab vision™
MS567B
200μg/mL Biotin 1mL Each for $1,581.78

MS567B0

 
thermo scientific™ lab vision™
MS567B0
200μg/mL Biotin 100μL Each for $362.57

MS567B1

 
thermo scientific™ lab vision™
MS567B1
200μg/mL Biotin 500μL Each for $997.41

MS567P

 
thermo scientific™ lab vision™
MS567P
200μg/mL Unlabeled 1mL Each for $1,265.24

MS567P0

 
thermo scientific™ lab vision™
MS567P0
200μg/mL Unlabeled 100μL Each for $270.71

MS567P1

 
thermo scientific™ lab vision™
MS567P1
200μg/mL Unlabeled 500μL Each for $764.94

MS567P1ABX

 
thermo scientific™ lab vision™
MS567P1ABX
1mg/mL Unlabeled 100μL Each for $842.78

MS567PABX

 
thermo scientific™ lab vision™
MS567PABX
1mg/mL Unlabeled 200μL Each for $1,366.71
Description & Specifications

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-1
Clone CA-4001 or CA719E3C
Applications Western Blot
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen N-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV
Isotype IgG1

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-2 (72kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody.

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids into an active form.of 62kDa.

Host Species: Mouse

Clone: CA-4001 or CA719E3C

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: N-terminal (APSPIIKFPGD-VAPKTDK)

Immunogen: N-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV

Molecular Weight: 72kDa (proform)

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Not suitable)
  • Inhibition of MMP-2 Activity

Note:

USA: RUO; Int'l: RUO