Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-13 (Collagenase-3) (Anti-Rat) Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) (Anti-Rat) Ab-3, Mouse Monoclonal Antibody.

$274.73 - $1,622.69

Specifications

Antigen MMP-13 (Collagenase-3) (Anti-Rat) Ab-3
Clone LIPCO IID1
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS827B

 
thermo scientific™ lab vision™
MS827B
200μg/mL Biotin 1mL Each for $1,622.69

MS827B0

 
thermo scientific™ lab vision™
MS827B0
200μg/mL Biotin 100µL Each for $372.16

MS827B1

 
thermo scientific™ lab vision™
MS827B1
200μg/mL Biotin 500µL Each for $1,012.43

MS827P

 
thermo scientific™ lab vision™
MS827P
200μg/mL Unlabeled 1mL Each for $1,300.45

MS827P0

 
thermo scientific™ lab vision™
MS827P0
200μg/mL Unlabeled 100µL Each for $274.73

MS827P1

 
thermo scientific™ lab vision™
MS827P1
200μg/mL Unlabeled 500µL Each for $778.10

MS827P1ABX

 
thermo scientific™ lab vision™
MS827P1ABX
1mg/mL Unlabeled 100µL Each for $864.65

MS827PABX

 
thermo scientific™ lab vision™
MS827PABX
1mg/mL Unlabeled 200µL Each for $1,413.35
Description & Specifications

Specifications

Antigen MMP-13 (Collagenase-3) (Anti-Rat) Ab-3
Clone LIPCO IID1
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen Rat MMP-13 from BC-1 cells
Isotype IgG2a kappa

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) (Anti-Rat) Ab-3, Mouse Monoclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Mouse

Clone: LIPCO IID1

Isotype: IgG1/κ

Species Reactivity: Rat and Mouse. Does not react with human. Others not known.

Epitope: Not determined

Immunogen: Rat MMP-13 from BC-1 cells

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) rat keratinocyte CCL-4 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO