Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-13 (Collagenase-3) (Anti-Rat) Ab-3, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) (Anti-Rat) Ab-3, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS827P1ABX

 View more versions of this product

Catalog No. MS827P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a kappa
Antigen MMP-13 (Collagenase-3) (Anti-Rat) Ab-3
Immunogen Rat MMP-13 from BC-1 cells
Quantity 100µL
Clone LIPCO IID1
Host Species Mouse
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) (Anti-Rat) Ab-3, Mouse Monoclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Mouse

Clone: LIPCO IID1

Isotype: IgG1/κ

Species Reactivity: Rat and Mouse. Does not react with human. Others not known.

Epitope: Not determined

Immunogen: Rat MMP-13 from BC-1 cells

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) rat keratinocyte CCL-4 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

  • Immunofluorescence

  • Immunoprecipitation (Native verified)
  • Note:

    USA: RUO; Int'l: RUO