Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody.

$271.47 - $1,285.06

Specifications

Antigen MMP-13 (Collagenase-3) Ab-5
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1681P

 
thermo scientific™ lab vision™
RB1681P
Purified Ab with BSA and Azide 1mL Each for $1,193.00

RB1681P0

 
thermo scientific™ lab vision™
RB1681P0
Purified Ab with BSA and Azide 100μL Each for $271.47

RB1681P1

 
thermo scientific™ lab vision™
RB1681P1
Purified Ab with BSA and Azide 500μL Each for $728.61

RB1681P1ABX

 
thermo scientific™ lab vision™
RB1681P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $801.71

RB1681PABX

 
thermo scientific™ lab vision™
RB1681PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,285.06
Description & Specifications

Specifications

Antigen MMP-13 (Collagenase-3) Ab-5
Concentration 1mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen Human recombinant MMP-13 protein
Isotype IgG

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant MMP-13 protein

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: CCL-4 medium

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO