Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1681PABX

 View more versions of this product

Catalog No. RB1681PABX

  • / Each
 

Description & Specifications

Specifications

Antigen MMP-13 (Collagenase-3) Ab-5
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen Human recombinant MMP-13 protein
Isotype IgG
Quantity 1mL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant MMP-13 protein

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: CCL-4 medium

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO