Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1681P1ABX

 View more versions of this product

Catalog No. RB1681P1ABX

  • / Each
 

Description & Specifications

Specifications

Immunogen Human recombinant MMP-13 protein
Cross Reactivity Human
Format Concentrated
Applications Western Blotting
Quantity 500µL
Regulatory Status RUO
Antigen MMP-13 (Collagenase-3) Ab-5
Isotype IgG
Host Species Rabbit
Classification Polyclonal
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Proteases And Inhibitors
Type Primary
Conjugate Unlabeled
Item Description 500uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Ensure accurate, reproducible results in experiments with the Thermo Scientific™ MMP-13 (Collagenase-3) Ab-5, Rabbit Polyclonal Antibody.

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant MMP-13 protein

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: CCL-4 medium

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO